This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01761260_m1
See other EEF2KMT GE Assays ›
Gene Symbol:
Eef2kmt
Gene Name:
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases:
Fam86a, RGD1304969
Chromosome Location:
Chr.10: 10530297 - 10540428 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01761260_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef2kmt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106975.1 NP_001100445.1 1 - 2 120 83
XM_006245785.2 XP_006245847.1 1 - 2 188 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000000119.rn.1

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-lysine trimethylation
protein-lysine N-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000000972.rn.1 TC1000000119.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000000973.rn.1 TC1000000119.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000000974.rn.1 TC1000000119.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0