This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01759754_g1
See other CABP4 GE Assays ›
Gene Symbol:
Cabp4
Gene Name:
calcium binding protein 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 219383451 - 219389314 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01759754_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108926.1 NP_001102396.1 5 - 6 844 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100007055.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

visual perception
phototransduction
biological_process
photoreceptor cell morphogenesis
retinal cone cell development
retinal bipolar neuron differentiation
molecular_function
calcium ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142904 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 734934 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 765141 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 61619 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 948956 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142904 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142904 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142904 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P114384 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10727455 - -
Affymetrix Rat Gene 1.0 ST Array 10727459 - -
Affymetrix Rat Gene 1.0 ST Array 10727454 - -
Affymetrix Rat Gene 1.0 ST Array 10727456 - -
Affymetrix Rat Gene 1.0 ST Array 10727458 - -
Affymetrix Rat Gene 1.0 ST Array 10727461 - -
Affymetrix Rat Gene 1.1 ST Array 10727453 - -
Rat Genome 230 2.0 Array 1385588_at - -
Rat Genome 230 2.0 Array 1385588_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100056050.rn.1 TC0100007055.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100056051.rn.1 TC0100007055.rn.1 RTA TC
Rat Expression Set 230 1385588_PM_at - -
Rat Expression Set 230 1385588_at - -
Affymetrix Rat Gene 1.0 ST Array 10727453 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21854164 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1355061 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0