This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01759291_g1
See other LOC103690097 GE Assays ›
Gene Symbol:
LOC103690097 Rgs18
Gene Name:
regulator of G-protein signaling 18-like
regulator of G-protein signaling 18
Gene Aliases:
-
-
Chromosome Location:
Chr.13: 61421718 - 61423679 on Build Rnor_6.0
Chromosome Location:
Chr.13: 61564381 - 61591139 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01759291_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC103690097
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008769583.1 XP_008767805.1 363 86
Gene Symbol: Rgs18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001047084.1 NP_001040549.1 1 - 2 114 86
GenBank mRNA AY651776.1 - 1 - 2 114 86

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300010738.rn.1 TC1300001560.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300010737.rn.1 TC1300001560.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0