This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01757867_g1
See other GPR98 GE Assays ›
Gene Symbol:
Gpr98 RGD1562101
Gene Name:
G protein-coupled receptor 98
similar to very large G-protein coupled receptor 1
Gene Aliases:
-
-
Chromosome Location:
-
Chromosome Location:
Chr.2: 8926843 - 9504474 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01757867_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA D87256.1 - 12 - 13 2467 96
Gene Symbol: RGD1562101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008760641.1 XP_008758863.1 12 - 13 2608 96

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Transporter 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
visual perception
sensory perception of sound
photoreceptor cell maintenance
maintenance of organ identity
inner ear development
neurological system process
detection of mechanical stimulus involved in sensory perception of sound
sensory perception of light stimulus
inner ear receptor stereocilium organization
G-protein coupled receptor activity
calcium ion binding
myosin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0