This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01756003_m1
See other ICE1 GE Assays ›
Gene Symbol:
Ice1
Gene Name:
interactor of little elongation complex ELL subunit 1
Gene Aliases:
RGD1309747
Chromosome Location:
Chr.1: 35280481 - 35322769 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01756003_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ice1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_225143.7 XP_225143.5 15 - 16 6298 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
positive regulation of protein complex assembly
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
positive regulation of transcription from RNA polymerase III promoter
positive regulation of intracellular protein transport
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0