This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01755210_m1
See other GJD4 GE Assays ›
Gene Symbol:
Gjd4
Gene Name:
gap junction protein, delta 4
Gene Aliases:
Cx39
Chromosome Location:
Chr.17: 62297086 - 62299655 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01755210_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gjd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100487.1 NP_001093957.1 1 - 2 65 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700002004.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s)

cell communication
regulation of satellite cell activation involved in skeletal muscle regeneration

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P153287 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153287 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153287 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P153287 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P180067 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10798950 - -
Affymetrix Rat Gene 1.1 ST Array 10798949 - -
Affymetrix Rat Clariom S Assay PSR1700013542.rn.1 TC1700002004.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700013543.rn.1 TC1700002004.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10798949 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0