This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01752189_g1
See other DNASE1L2 GE Assays ›
Gene Symbol:
Dnase1l2
Gene Name:
deoxyribonuclease I-like 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 13812335 - 13814512 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01752189_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnase1l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001057412.3 XP_001057412.4 4 - 5 626 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA catabolic process, endonucleolytic
hair follicle development
corneocyte development
DNA catabolic process
endodeoxyribonuclease activity
endodeoxyribonuclease activity, producing 5'-phosphomonoesters

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0