This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01751753_g1
See other STUB1 GE Assays ›
Gene Symbol:
Stub1
Gene Name:
STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase
Gene Aliases:
Chip
Chromosome Location:
Chr.10: 15197754 - 15200035 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01751753_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stub1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025625.2 NP_001020796.2 1 - 2 266 68
GenBank mRNA BC099240.1 - 1 - 2 7445727 68
CB568781.1 - 1 - 2 267 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000002441.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stub1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
ubiquitin-dependent SMAD protein catabolic process
endoplasmic reticulum unfolded protein response
positive regulation of protein ubiquitination
regulation of glucocorticoid metabolic process
negative regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of ubiquitin-protein transferase activity
protein autoubiquitination
protein K63-linked ubiquitination
cellular response to misfolded protein
positive regulation of chaperone-mediated protein complex assembly
G-protein coupled receptor binding
ubiquitin-protein transferase activity
protein binding
ligase activity
enzyme binding
kinase binding
Hsp70 protein binding
TPR domain binding
ubiquitin protein ligase binding
ubiquitin-ubiquitin ligase activity
protein homodimerization activity
SMAD binding
misfolded protein binding
Hsp90 protein binding
ubiquitin protein ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000022007.rn.1 TC1000002441.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000022006.rn.1 TC1000002441.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000022003.rn.1 TC1000002441.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000022009.rn.1 TC1000002441.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0