This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01750706_m1
See other MRPL45 GE Assays ›
Gene Symbol:
Mrpl45
Gene Name:
mitochondrial ribosomal protein L45
Gene Aliases:
-
Chromosome Location:
Chr.10: 85257876 - 85269806 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01750706_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl45
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105834.1 NP_001099304.1 1 - 2 118 117

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001105834.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000013638.rn.1 TC1000001539.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000013637.rn.1 TC1000001539.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000013639.rn.1 TC1000001539.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0