This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01747806_m1
See other GPR132 GE Assays ›
Gene Symbol:
Gpr132
Gene Name:
G protein-coupled receptor 132
Gene Aliases:
G2a
Chromosome Location:
Chr.6: 137652213 - 137670156 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01747806_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170595.1 NP_001164066.1 1 - 2 83 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001170595.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600022961.rn.1 TC0600003254.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600022960.rn.1 TC0600003254.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600022959.rn.1 TC0600003254.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0