This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01648310_m1
See other TMEM132D GE Assays ›
Gene Symbol:
Tmem132d
Gene Name:
transmembrane protein 132D
Gene Aliases:
MOLT
Chromosome Location:
Chr.12: 31933716 - 32608616 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01648310_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem132d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198727.1 NP_942022.1 5 - 6 1480 87
XM_006249266.2 XP_006249328.2 5 - 6 2106 87
GenBank mRNA AB100356.1 - 5 - 6 1480 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000545.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem132d

Gene Ontology Categories:

Function(s) Process(es)

biological_process
negative regulation of phosphatase activity
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200004661.rn.1 TC1200000545.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0