This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01647218_m1
See other SDS GE Assays ›
Gene Symbol:
Sds
Gene Name:
serine dehydratase
Gene Aliases:
RATSDHE1, SDH2, Sdh, Sdhe1, TDH
Chromosome Location:
Chr.12: 41620429 - 41627741 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01647218_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sds
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053962.3 NP_446414.3 2 - 3 129 106
XM_008769213.1 XP_008767435.1 1 - 2 627 106
GenBank mRNA BC088110.1 - 2 - 3 129 7323391 106
J03863.1 - 2 - 3 219 106
Y00752.1 - 2 - 3 146 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200001646.rn.1

Additional Information:

This assay is in the 5'UTR region of the BC088110.1,J03863.1,NM_053962.3,XM_008769213.1,Y00752.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sds

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
L-serine catabolic process
threonine catabolic process
response to nutrient levels
response to cobalamin
pyruvate biosynthetic process
response to amino acid
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
pyridoxal phosphate binding
protein homodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200014176.rn.1 TC1200001646.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0