This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01644169_m1
See other GPR64 GE Assays ›
Gene Symbol:
Gpr64
Gene Name:
G protein-coupled receptor 64
Gene Aliases:
He6, Re6
Chromosome Location:
Chr.X: 36930186 - 37054925 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01644169_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr64
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270871.1 NP_001257800.1 24 - 25 2705 148
NM_001270872.1 NP_001257801.1 23 - 24 2669 148
NM_181366.2 NP_852031.1 25 - 26 2753 148
XM_006256916.2 XP_006256978.1 25 - 26 2793 148
XM_008773154.1 XP_008771376.1 24 - 25 3133 148
XM_008773155.1 XP_008771377.1 24 - 25 2743 148
XM_008773156.1 XP_008771378.1 23 - 24 3097 148
XM_008773157.1 XP_008771379.1 23 - 24 3091 148
XM_008773158.1 XP_008771380.1 22 - 23 3055 148
XM_008773159.1 XP_008771381.1 21 - 22 2646 148
GenBank mRNA AF538953.1 - 25 - 26 2763 148
AF538958.1 - 24 - 25 2715 148
AF538959.1 - 23 - 24 2679 148

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0X00001687.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr64

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00010919.rn.1 TC0X00001687.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0X00010918.rn.1 TC0X00001687.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0