This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01535808_m1
See other LOC100910882 GE Assays ›
Gene Symbol:
LOC100910882 Rbm39
Gene Name:
RNA-binding protein 39-like
RNA binding motif protein 39
Gene Aliases:
-
Rnp1, Rnpc2, Rrm
Chromosome Location:
Chr.3: 152226018 - 152259228 on Build Rnor_6.0
Chromosome Location:
Chr.3: 151691505 - 151724715 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01535808_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100910882
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006235446.2 XP_006235508.1 3 - 4 708 75
XM_008762418.1 XP_008760640.1 2 - 3 2198 75
Gene Symbol: Rbm39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006235335.2 XP_006235397.1 708 75
XM_008762352.1 XP_008760574.1 2 - 3 2198 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006235335.2, XM_006235446.2, XM_008762352.1, XM_008762418.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
nucleotide binding
RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0