This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01535063_m1
See other GULO GE Assays ›
Gene Symbol:
Gulo
Gene Name:
gulonolactone (L-) oxidase
Gene Aliases:
-
Chromosome Location:
Chr.15: 42671206 - 42693711 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01535063_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gulo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022220.2 NP_071556.2 5 - 6 458 118
XM_006252146.2 XP_006252208.1 5 - 6 599 118
GenBank mRNA BC089803.1 - 5 - 6 458 7324344 118
FQ219140.1 - 5 - 6 483 118
FQ219632.1 - 5 - 6 566 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1500001853.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gulo

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

L-ascorbic acid biosynthetic process
oxidation-reduction process
D-arabinono-1,4-lactone oxidase activity
UDP-N-acetylmuramate dehydrogenase activity
L-gulonolactone oxidase activity
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1500012422.rn.1 TC1500001853.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm