This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01534136_g1
See other NHLH2 GE Assays ›
Gene Symbol:
Nhlh2
Gene Name:
nescient helix loop helix 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 204427577 - 204432946 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01534136_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nhlh2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106457.1 NP_001099927.1 1 - 2 181 149

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200001859.rn.1

Additional Information:

This assay is in the 5'UTR region of the NM_001106457.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

mating behavior
biological_process
ovulation cycle
RNA polymerase II activating transcription factor binding
molecular_function
protein dimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157873 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157868 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 638144 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 343986 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 271394 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 442587 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 700188 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157873 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157868 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157873 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157868 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157868 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157873 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P473345 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10817880 - -
Affymetrix Rat Gene 1.1 ST Array 10817879 - -
Affymetrix Rat Clariom S Assay PSR0200012626.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012630.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012631.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012623.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012627.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012632.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012622.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012624.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200012629.rn.1 TC0200001859.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10817879 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21321662 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1368198 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm