This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01533623_m1
See other GPR89B GE Assays ›
Gene Symbol:
Gpr89b
Gene Name:
G protein-coupled receptor 89B
Gene Aliases:
Gpr89, RGD1304837
Chromosome Location:
Chr.2: 198999945 - 199038702 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01533623_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr89b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001139486.1 NP_001132958.1 11 - 12 1103 111
XM_006232976.2 XP_006233038.1 9 - 10 1092 111
GenBank mRNA AY724489.1 - 11 - 12 1034 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200004267.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
skin development
regulation of anion transport
intracellular pH reduction
signal transducer activity
voltage-gated anion channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200029307.rn.1 TC0200004267.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200029306.rn.1 TC0200004267.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0