This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01531770_m1
See other GPR56 GE Assays ›
Gene Symbol:
Gpr56
Gene Name:
G protein-coupled receptor 56
Gene Aliases:
-
Chromosome Location:
Chr.19: 10423534 - 10460716 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01531770_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152242.2 NP_689448.2 12 - 13 1955 77
XM_006255066.2 XP_006255128.1 12 - 13 3007 77
XM_008772264.1 XP_008770486.1 12 - 13 2157 77
XM_008772265.1 XP_008770487.1 13 - 14 2421 77
XM_008772266.1 XP_008770488.1 12 - 13 3667 77
XM_008772267.1 XP_008770489.1 12 - 13 3930 77
XM_008772268.1 XP_008770490.1 13 - 14 2254 77
XM_008772269.1 XP_008770491.1 12 - 13 3036 77
XM_008772270.1 XP_008770492.1 12 - 13 3512 77
GenBank mRNA AF529886.1 - 12 - 13 1956 77
CK482497.1 - 2 - 3 151 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900000985.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr56

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cell adhesion
G-protein coupled receptor signaling pathway
Rho protein signal transduction
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
extracellular matrix binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P167965 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095830 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 457598 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 41340 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 746313 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 907827 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 608961 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 76535 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 609635 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 818121 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 634901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 223149 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 544134 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 595446 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 544049 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 277025 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 309626 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 456491 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 755150 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 745215 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 248329 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 811456 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 319826 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 746834 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 465938 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 371638 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 746768 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 521595 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 864413 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 23152 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 164407 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 170909 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 26300 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095830 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P167965 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P167965 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095830 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P167965 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095830 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P167965 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 74785 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 52238 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10809230 - -
Affymetrix Rat Gene 1.0 ST Array 10809220 - -
Affymetrix Rat Gene 1.0 ST Array 10809223 - -
Affymetrix Rat Gene 1.0 ST Array 10809221 - -
Affymetrix Rat Gene 1.0 ST Array 10809225 - -
Affymetrix Rat Gene 1.0 ST Array 10809231 - -
Affymetrix Rat Gene 1.0 ST Array 10809219 - -
Affymetrix Rat Gene 1.0 ST Array 10809229 - -
Affymetrix Rat Gene 1.0 ST Array 10809227 - -
Affymetrix Rat Gene 1.0 ST Array 10809222 - -
Affymetrix Rat Gene 1.0 ST Array 10809226 - -
Affymetrix Rat Gene 1.0 ST Array 10809228 - -
Affymetrix Rat Gene 1.0 ST Array 10809224 - -
Affymetrix Rat Gene 1.0 ST Array 10809217 - -
Affymetrix Rat Gene 1.0 ST Array 10809218 - -
Affymetrix Rat Gene 1.1 ST Array 10809216 - -
Rat Genome 230 2.0 Array 1371696_PM_at - -
Rat Genome 230 2.0 Array 1371696_at - -
Affymetrix HT Rat Focus Array 1371696_at - -
Affymetrix HT Rat Focus Array 1371696_PM_at - -
Affymetrix Rat Clariom S Assay PSR1900007804.rn.1 TC1900000985.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900007806.rn.1 TC1900000985.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900007805.rn.1 TC1900000985.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900007803.rn.1 TC1900000985.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900007808.rn.1 TC1900000985.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900007807.rn.1 TC1900000985.rn.1 RTA TC
Rat Expression Set 230 1371696_PM_at - -
Rat Expression Set 230 1371696_at - -
Affymetrix Rat Gene 1.0 ST Array 10809216 - -
Rat Genome U34 Set rc_AA819206_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20925841 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1371270 - -
RatRef-12 Expression BeadChip ILMN_53094 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0