This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01524230_m1
See other LOC683722 GE Assays ›
Gene Symbol:
LOC683722
Gene Name:
Fgfr1 oncogene partner
Gene Aliases:
Fgfr1op
Chromosome Location:
Chr.1: 53122330 - 53153108 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01524230_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC683722
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101008.1 NP_001094478.1 6 - 7 602 93
XM_008758772.1 XP_008756994.1 6 - 7 1013 93
GenBank mRNA BC098026.1 - 6 - 7 602 7455942 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100004508.rn.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
positive regulation of cell proliferation
positive regulation of cell growth
positive regulation of cell migration
microtubule anchoring
negative regulation of protein tyrosine kinase activity
protein kinase binding
protein tyrosine kinase inhibitor activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100035759.rn.1 TC0100004508.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0