This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01523293_m1
See other ACADM GE Assays ›
Gene Symbol:
Acadm
Gene Name:
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases:
MCAD
Chromosome Location:
Chr.2: 260124418 - 260148589 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01523293_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acadm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016986.2 NP_058682.2 9 - 10 956 112
GenBank mRNA FQ214755.1 - 3 - 4 284 112
J02791.1 - 9 - 10 871 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200004932.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acadm

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

liver development
glycogen biosynthetic process
organic acid metabolic process
regulation of gluconeogenesis
fatty acid beta-oxidation
heart development
response to nutrient
response to cold
carnitine metabolic process
response to hormone
post-embryonic development
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to drug
response to starvation
carnitine biosynthetic process
response to copper ion
protein homotetramerization
response to glucocorticoid
medium-chain fatty acid metabolic process
medium-chain fatty acid catabolic process
cardiac muscle cell differentiation
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
isomerase activity
identical protein binding
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200034126.rn.1 TC0200004932.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0