This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01520971_m1
See other GPR137B GE Assays ›
Gene Symbol:
Gpr137b
Gene Name:
G protein-coupled receptor 137B
Gene Aliases:
Tm7sf1
Chromosome Location:
Chr.17: 90670795 - 90709003 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01520971_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr137b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105978.1 NP_001099448.1 2 - 3 291 93
XM_006254372.2 XP_006254434.1 3 - 4 793 93
XM_006254373.2 XP_006254435.1 3 - 4 788 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700001148.rn.1

Additional Information:

This assay is in the 5'UTR region of the NM_001105978.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700007952.rn.1 TC1700001148.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700007953.rn.1 TC1700001148.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700007951.rn.1 TC1700001148.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700007950.rn.1 TC1700001148.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0