This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01517952_m1
See other GPR68 GE Assays ›
Gene Symbol:
Gpr68
Gene Name:
G protein-coupled receptor 68
Gene Aliases:
-
Chromosome Location:
Chr.6: 124874151 - 124903949 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01517952_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108049.1 NP_001101519.1 556 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001108049.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Inventoried

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
positive regulation of insulin secretion
positive regulation of insulin secretion involved in cellular response to glucose stimulus
negative regulation of monocyte differentiation
cellular response to pH
positive regulation of osteoclast development
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0