This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01515608_m1
See other EPB4.1 GE Assays ›
Gene Symbol:
Epb4.1
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
RGD1564762
Chromosome Location:
Chr.5: 150081727 - 150243186 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01515608_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Epb4.1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_006239085.2
XP_006239147.1
3 - 4
984
69
XM_006239086.2
XP_006239148.1
3 - 4
984
69
XM_006239087.2
XP_006239149.1
3 - 4
2936
69
XM_006239088.2
XP_006239150.1
3 - 4
984
69
XM_006239089.2
XP_006239151.1
3 - 4
984
69
XM_006239090.2
XP_006239152.1
3 - 4
984
69
XM_006239091.2
XP_006239153.1
3 - 4
984
69
XM_006239092.2
XP_006239154.1
3 - 4
985
69
XM_006239093.2
XP_006239155.1
3 - 4
820
69
XM_006239094.2
XP_006239156.1
3 - 4
984
69
XM_006239095.1
XP_006239157.1
3 - 4
843
69
XM_006239096.2
XP_006239158.1
3 - 4
982
69
XM_006239097.2
XP_006239159.1
3 - 4
982
69
XM_006239098.2
XP_006239160.1
3 - 4
982
69
XM_006239099.2
XP_006239161.1
3 - 4
982
69
XM_006239100.2
XP_006239162.1
3 - 4
981
69
XM_006239101.2
XP_006239163.1
3 - 4
983
69
XM_006239103.2
XP_006239165.1
3 - 4
981
69
XM_006239104.2
XP_006239166.1
3 - 4
982
69
XM_006239105.2
XP_006239167.1
3 - 4
984
69
XM_008764184.1
XP_008762406.1
3 - 4
981
69
XM_008764185.1
XP_008762407.1
3 - 4
981
69
XM_232771.8
XP_232771.6
3 - 4
820
69
GenBank mRNA
FQ233548.1
-
3 - 4
779
69
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Rattus norvegicus
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural molecule activity
protein binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0