This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01515607_m1
See other EPB4.1 GE Assays ›
Gene Symbol:
Epb4.1
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
RGD1564762
Chromosome Location:
Chr.5: 150081727 - 150243186 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01515607_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006239085.2 XP_006239147.1 2 - 3 775 75
XM_006239086.2 XP_006239148.1 2 - 3 775 75
XM_006239087.2 XP_006239149.1 2 - 3 2727 75
XM_006239088.2 XP_006239150.1 2 - 3 775 75
XM_006239089.2 XP_006239151.1 2 - 3 775 75
XM_006239090.2 XP_006239152.1 2 - 3 775 75
XM_006239091.2 XP_006239153.1 2 - 3 775 75
XM_006239092.2 XP_006239154.1 2 - 3 776 75
XM_006239093.2 XP_006239155.1 2 - 3 611 75
XM_006239094.2 XP_006239156.1 2 - 3 775 75
XM_006239095.1 XP_006239157.1 2 - 3 634 75
XM_006239096.2 XP_006239158.1 2 - 3 773 75
XM_006239097.2 XP_006239159.1 2 - 3 773 75
XM_006239098.2 XP_006239160.1 2 - 3 773 75
XM_006239099.2 XP_006239161.1 2 - 3 773 75
XM_006239100.2 XP_006239162.1 2 - 3 772 75
XM_006239101.2 XP_006239163.1 2 - 3 774 75
XM_006239103.2 XP_006239165.1 2 - 3 772 75
XM_006239104.2 XP_006239166.1 2 - 3 773 75
XM_006239105.2 XP_006239167.1 2 - 3 775 75
XM_008764184.1 XP_008762406.1 2 - 3 772 75
XM_008764185.1 XP_008762407.1 2 - 3 772 75
XM_232771.8 XP_232771.6 2 - 3 611 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006239095.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural molecule activity
protein binding
1-phosphatidylinositol binding
spectrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0