Assay Details
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239085.2 | XP_006239147.1 | 19 - 20 | 3094 | 65 | |
XM_006239086.2 | XP_006239148.1 | 19 - 20 | 3079 | 65 | ||
XM_006239087.2 | XP_006239149.1 | 19 - 20 | 5046 | 65 | ||
XM_006239088.2 | XP_006239150.1 | 19 - 20 | 3055 | 65 | ||
XM_006239089.2 | XP_006239151.1 | 18 - 19 | 3052 | 65 | ||
XM_006239090.2 | XP_006239152.1 | 18 - 19 | 3043 | 65 | ||
XM_006239091.2 | XP_006239153.1 | 17 - 18 | 2995 | 65 | ||
XM_006239093.2 | XP_006239155.1 | 19 - 20 | 2930 | 65 | ||
XM_006239094.2 | XP_006239156.1 | 16 - 17 | 2932 | 65 | ||
XM_006239095.1 | XP_006239157.1 | 19 - 20 | 2953 | 65 | ||
XM_006239096.2 | XP_006239158.1 | 18 - 19 | 2633 | 65 | ||
XM_006239097.2 | XP_006239159.1 | 18 - 19 | 2618 | 65 | ||
XM_006239098.2 | XP_006239160.1 | 17 - 18 | 2582 | 65 | ||
XM_006239099.2 | XP_006239161.1 | 16 - 17 | 2540 | 65 | ||
XM_006239100.2 | XP_006239162.1 | 16 - 17 | 2518 | 65 | ||
XM_006239103.2 | XP_006239165.1 | 14 - 15 | 2419 | 65 | ||
XM_008764184.1 | XP_008762406.1 | 15 - 16 | 2482 | 65 | ||
XM_008764185.1 | XP_008762407.1 | 14 - 15 | 2416 | 65 | ||
XM_232771.8 | XP_232771.6 | 15 - 16 | 2321 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
