This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01513019_g1
See other ITPA GE Assays ›
Gene Symbol:
Itpa
Gene Name:
inosine triphosphatase (nucleoside triphosphate pyrophosphatase)
Gene Aliases:
-
Chromosome Location:
Chr.3: 123209611 - 123221266 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01513019_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Itpa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107774.1 NP_001101244.1 3 - 4 253 84
GenBank mRNA FQ230286.1 - 3 - 4 216 84
FQ231907.1 - 3 - 4 216 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300001730.rn.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

nucleotide phosphatase

Gene Ontology Categories:

Function(s) Process(es)

ITP catabolic process
biological_process
deoxyribonucleoside triphosphate catabolic process
chromosome organization
nucleotide binding
molecular_function
dITP diphosphatase activity
ITP diphosphatase activity
XTP diphosphatase activity
metal ion binding
nucleoside-triphosphate diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300012469.rn.1 TC0300001730.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0