This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01512179_m1
See other OPN5 GE Assays ›
Gene Symbol:
Opn5
Gene Name:
opsin 5
Gene Aliases:
Gpr136
Chromosome Location:
Chr.9: 21045457 - 21092517 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01512179_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Opn5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181772.1 NP_861437.1 3 - 4 419 74
GenBank mRNA AY288431.1 - 3 - 4 419 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900000306.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Opn5

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
protein-chromophore linkage
G-protein coupled receptor activity
11-cis retinal binding
photoreceptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112019 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 357016 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 577891 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 337197 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 122760 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 806412 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 655672 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 170777 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 304867 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 883495 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 362516 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 759524 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 66628 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 733391 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 49337 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 641035 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 410352 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 807940 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 548142 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 87333 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 644452 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 118651 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 140490 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36279 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 583407 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 432853 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 390569 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 177234 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 185184 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 129132 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 404055 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 693463 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 227405 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 148811 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 23532 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 63437 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 165721 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 66684 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 50316 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112019 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112019 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112019 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P294642 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 38782 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 6934 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 97465 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10921993 - -
Affymetrix Rat Gene 1.0 ST Array 10921999 - -
Affymetrix Rat Gene 1.0 ST Array 10921992 - -
Affymetrix Rat Gene 1.0 ST Array 10922001 - -
Affymetrix Rat Gene 1.0 ST Array 10921998 - -
Affymetrix Rat Gene 1.0 ST Array 10921996 - -
Affymetrix Rat Gene 1.0 ST Array 10922000 - -
Affymetrix Rat Gene 1.0 ST Array 10921991 - -
Affymetrix Rat Gene 1.0 ST Array 10921994 - -
Affymetrix Rat Gene 1.0 ST Array 10921995 - -
Affymetrix Rat Gene 1.1 ST Array 10921990 - -
Affymetrix Rat Clariom S Assay PSR0900002770.rn.1 TC0900000306.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002772.rn.1 TC0900000306.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002768.rn.1 TC0900000306.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002774.rn.1 TC0900000306.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002769.rn.1 TC0900000306.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002773.rn.1 TC0900000306.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10921990 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20796750 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1352793 - -
RatRef-12 Expression BeadChip ILMN_63081 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0