This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01511885_m1
See other GPR116 GE Assays ›
Gene Symbol:
Gpr116
Gene Name:
G protein-coupled receptor 116
Gene Aliases:
Gprhep, Ig-Hepta
Chromosome Location:
Chr.9: 20091121 - 20195708 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01511885_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr116
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139110.1 NP_620810.1 15 - 16 2380 100
XM_006244594.2 XP_006244656.1 16 - 17 2715 100
XM_006244597.2 XP_006244659.1 15 - 16 2594 100
XM_008766832.1 XP_008765054.1 17 - 18 2783 100
XM_008766833.1 XP_008765055.1 17 - 18 2708 100
XM_008766834.1 XP_008765056.1 17 - 18 2784 100
XM_008766835.1 XP_008765057.1 16 - 17 2662 100
GenBank mRNA AB019120.1 - 15 - 16 2380 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900001656.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr116

Gene Ontology Categories:

Function(s) Process(es)

energy reserve metabolic process
G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
glucose homeostasis
surfactant homeostasis
fat cell differentiation
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0900013627.rn.1 TC0900001656.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900013628.rn.1 TC0900001656.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0