This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01511670_g1
See other LOC100909464 GE Assays ›
Gene Symbol:
LOC100909464 Ppp2r5d
Gene Name:
serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform-like
protein phosphatase 2, regulatory subunit B', delta
Gene Aliases:
-
-
Chromosome Location:
Chr.9: 603900 - 606053 on Build Rnor_6.0
Chromosome Location:
Chr.9: 16580985 - 16610429 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01511670_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100909464
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003750611.3 XP_003750659.3 3 - 4 535 105
Gene Symbol: Ppp2r5d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001062510.5 XP_001062510.4 6 - 7 1401 105
XM_006244570.2 XP_006244632.1 6 - 7 1404 105
GenBank mRNA BC099800.1 - 5 - 6 693 105

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of catalytic activity
protein phosphatase type 2A regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0