This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01510110_m1
See other FOLH1 GE Assays ›
Gene Symbol:
Folh1
Gene Name:
folate hydrolase 1
Gene Aliases:
Naalad
Chromosome Location:
Chr.1: 150323768 - 150395415 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01510110_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Folh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_057185.2 NP_476533.1 6 - 7 896 104
GenBank mRNA AF040256.1 - 6 - 7 902 104
AF513486.2 - 6 - 7 896 104
U75973.1 - 6 - 7 853 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006006.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Folh1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
folic acid-containing compound metabolic process
carboxypeptidase activity
peptidase activity
metallopeptidase activity
dipeptidase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100047767.rn.1 TC0100006006.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100047766.rn.1 TC0100006006.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0