This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01509008_m1
See other LOC103690122 GE Assays ›
Gene Symbol:
LOC103690122 Ate1
Gene Name:
arginyl-tRNA--protein transferase 1-like
arginyltransferase 1
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 201024651 - 201083414 on Build Rnor_6.0
Chromosome Location:
Chr.1: 200810756 - 200893744 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01509008_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC103690122
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008759941.1 XP_008758163.1 1 - 2 186 106
XM_008759942.1 XP_008758164.1 1 - 2 186 106
XM_008759944.1 XP_008758166.1 1 - 2 187 106
XM_008759945.1 XP_008758167.1 1 - 2 186 106
Gene Symbol: Ate1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106300.1 NP_001099770.1 168 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006754.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

protein arginylation
arginyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100053360.rn.1 TC0100006754.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100053361.rn.1 TC0100006754.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0