This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01506637_g1
See other HSD3B7 GE Assays ›
Gene Symbol:
Hsd3b7
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Gene Aliases:
Cca2
Chromosome Location:
Chr.1: 199248084 - 199251747 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01506637_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd3b7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139329.1 NP_647545.1 1 - 2 102 53
XM_006230212.2 XP_006230274.1 1 - 2 488 53
GenBank mRNA AB000199.1 - 1 - 2 102 53

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100002660.rn.1

Additional Information:

This assay is in the 5'UTR region of the AB000199.1,NM_139329.1,XM_006230212.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsd3b7

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
steroid biosynthetic process
bile acid metabolic process
oxidation-reduction process
3-beta-hydroxy-delta5-steroid dehydrogenase activity
isomerase activity
cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100020666.rn.1 TC0100002660.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100020665.rn.1 TC0100002660.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100020669.rn.1 TC0100002660.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0