Assay Details
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130571.1 | NP_001124043.1 | 25 - 26 | 3092 | 60 | |
XM_006241853.2 | XP_006241915.1 | 25 - 26 | 3093 | 60 | ||
GenBank mRNA | BC168713.1 | - | 25 - 26 | 3006 | 8367087 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
