This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01501459_g1
See other LOC100911692 GE Assays ›
Gene Symbol:
LOC100911692 Cracr2b
Gene Name:
EF-hand calcium-binding domain-containing protein 4A-like
calcium release activated channel regulator 2B
Gene Aliases:
-
Efcab4a, RGD1560911
Chromosome Location:
Chr.1: 214002860 - 214008970 on Build Rnor_6.0
Chromosome Location:
Chr.1: 214439766 - 214445876 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01501459_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100911692
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006230595.2 XP_006230657.1 9 - 10 2570 102
XM_006230596.2 XP_006230658.1 10 - 11 1934 102
XM_006230597.2 XP_006230659.1 9 - 10 1822 102
Gene Symbol: Cracr2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127541.1 NP_001121013.1 1197 102
XM_006230559.2 XP_006230621.1 9 - 10 2570 102
XM_006230561.2 XP_006230623.1 9 - 10 1822 102
GenBank mRNA BC158864.1 - 1197 7936496 102

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
regulation of store-operated calcium entry
calcium ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P389178 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 153258 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 930057 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 362160 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 491401 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 323135 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 130244 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7948 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389178 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389178 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P389178 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389178 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 9031 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 2441 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 47626 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10712337 - -
Affymetrix Rat Gene 1.0 ST Array 10712331 - -
Affymetrix Rat Gene 1.0 ST Array 10712339 - -
Affymetrix Rat Gene 1.0 ST Array 10712336 - -
Affymetrix Rat Gene 1.0 ST Array 10712332 - -
Affymetrix Rat Gene 1.0 ST Array 10712334 - -
Affymetrix Rat Gene 1.0 ST Array 10712338 - -
Affymetrix Rat Gene 1.0 ST Array 10712335 - -
Affymetrix Rat Gene 1.0 ST Array 10712333 - -
Affymetrix Rat Gene 1.1 ST Array 10712330 - -
Rat Genome 230 2.0 Array 1390285_at - -
Rat Genome 230 2.0 Array 1390285_PM_at - -
Affymetrix HT Rat Focus Array 1390285_at - -
Affymetrix HT Rat Focus Array 1390285_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100022781.rn.1 TC0100002881.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100022782.rn.1 TC0100002881.rn.1 RTA TC
Rat Expression Set 230 1390285_PM_at - -
Rat Expression Set 230 1390285_at - -
Affymetrix Rat Gene 1.0 ST Array 10712330 - -
Rat Genome U34 Set rc_AI178948_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20799257 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1351563 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0