This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01501456_g1
See other CRACR2B GE Assays ›
Gene Symbol:
Cracr2b LOC100911692
Gene Name:
calcium release activated channel regulator 2B
EF-hand calcium-binding domain-containing protein 4A-like
Gene Aliases:
Efcab4a, RGD1560911
-
Chromosome Location:
Chr.1: 214439766 - 214445876 on Build Rnor_6.0
Chromosome Location:
Chr.1: 214002860 - 214008970 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01501456_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cracr2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127541.1 NP_001121013.1 768 151
XM_006230559.2 XP_006230621.1 5 - 6 2141 151
XM_006230561.2 XP_006230623.1 5 - 6 1393 151
GenBank mRNA BC158864.1 - 768 7936496 151
Gene Symbol: LOC100911692
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006230595.2 XP_006230657.1 5 - 6 2141 151
XM_006230596.2 XP_006230658.1 6 - 7 1505 151
XM_006230597.2 XP_006230659.1 5 - 6 1393 151

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
regulation of store-operated calcium entry
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100022777.rn.1 TC0100002881.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100022778.rn.1 TC0100002881.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0