This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01492963_m1
See other SLC2A3 GE Assays ›
Gene Symbol:
Slc2a3 LOC100909595
Gene Name:
solute carrier family 2 (facilitated glucose transporter), member 3
solute carrier family 2, facilitated glucose transporter member 3-like
Gene Aliases:
GLUT3
-
Chromosome Location:
Chr.4: 155549991 - 155626030 on Build Rnor_6.0
Chromosome Location:
Chr.4: 155750318 - 155763834 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01492963_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc2a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017102.2 NP_058798.2 1071 81
XM_006237295.1 XP_006237357.1 1299 81
XM_006237296.2 XP_006237358.1 1618 81
XM_006237297.1 XP_006237359.1 1303 81
XM_006237298.2 XP_006237360.1 1581 81
GenBank mRNA D13962.1 - 1155 81
U17978.1 - 1071 81
Gene Symbol: LOC100909595
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003749833.3 XP_003749881.1 8 - 9 1405 81

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc2a3
Click here to find siRNAs for Gene LOC100909595

Gene Ontology Categories:

Function(s) Process(es)

response to glucose
monosaccharide transport
glucose transport
hexose transmembrane transport
negative regulation of apoptotic process
glucose import
transmembrane transport
dehydroascorbic acid transport
glucose transmembrane transporter activity
protein binding
glucose binding
monosaccharide transmembrane transporter activity
galactoside binding
kinase binding
xylose binding
dehydroascorbic acid transporter activity
D-glucose transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0400026101.rn.1 TC0400003694.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header