This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01491715_m1
See other ZFP532 GE Assays ›
Gene Symbol:
Zfp532
Gene Name:
zinc finger protein 532
Gene Aliases:
Znf532
Chromosome Location:
Chr.18: 61257522 - 61368371 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01491715_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp532
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107382.1 NP_001100852.1 7 - 8 3310 118
XM_006254865.2 XP_006254927.1 7 - 8 5600 118
XM_006254866.2 XP_006254928.1 7 - 8 5039 118
XM_006254868.2 XP_006254930.1 7 - 8 3627 118
XM_006254869.2 XP_006254931.1 7 - 8 3472 118
XM_006254870.2 XP_006254932.1 7 - 8 5004 118
XM_006254871.2 XP_006254933.1 6 - 7 3185 118
XM_006254872.2 XP_006254934.1 7 - 8 4384 118
XM_006254873.2 XP_006254935.1 6 - 7 5490 118
XM_006254875.2 XP_006254937.1 3 - 4 3704 118
XM_008772142.1 XP_008770364.1 6 - 7 4226 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800000699.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Process(es)

nucleic acid binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0