This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01488425_m1
See other CNOT7 GE Assays ›
Gene Symbol:
Cnot7
Gene Name:
CCR4-NOT transcription complex, subunit 7
Gene Aliases:
-
Chromosome Location:
Chr.16: 54899452 - 54918616 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01488425_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnot7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107313.1 NP_001100783.1 5 - 6 914 133
XM_008771290.1 XP_008769512.1 5 - 6 1087 133
XM_008771291.1 XP_008769513.1 5 - 6 1057 133
XM_008771292.1 XP_008769514.1 5 - 6 957 133
GenBank mRNA BC167766.1 - 5 - 6 873 7455944 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1600000634.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
nuclear-transcribed mRNA poly(A) tail shortening
deadenylation-dependent decapping of nuclear-transcribed mRNA
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of gene expression
gene silencing by RNA
cytoplasmic mRNA processing body assembly
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
positive regulation of transcription from RNA polymerase II promoter
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
positive regulation of mRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
3'-5'-exoribonuclease activity
nucleic acid binding
exoribonuclease activity
poly(A)-specific ribonuclease activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1600005062.rn.1 TC1600000634.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0