This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01486500_g1
See other EIF3G GE Assays ›
Gene Symbol:
Eif3g
Gene Name:
eukaryotic translation initiation factor 3, subunit G
Gene Aliases:
Eif3s4
Chromosome Location:
Chr.8: 21911632 - 21915664 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01486500_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013095.1 NP_001013113.1 4 - 5 249 73
GenBank mRNA BC086383.1 - 4 - 5 249 7301572 73
FQ213005.1 - 4 - 5 290 73
FQ225883.1 - 4 - 5 282 73
FQ226276.1 - 4 - 5 281 73
FQ229703.1 - 4 - 5 280 73
FQ232595.1 - 4 - 5 279 73
FQ232661.1 - 4 - 5 279 73
FQ234018.1 - 4 - 5 282 73
FQ234727.1 - 2 - 3 131 73
FQ234921.1 - 4 - 5 280 73
FQ234929.1 - 4 - 5 282 73
FQ235264.1 - 1 - 2 63 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800002053.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3g

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
translational initiation
regulation of translational initiation
nucleotide binding
translation initiation factor activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800016410.rn.1 TC0800002053.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0