This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01483613_m1
See other RFWD2 GE Assays ›
Gene Symbol:
Rfwd2
Gene Name:
ring finger and WD repeat domain 2, E3 ubiquitin protein ligase
Gene Aliases:
Cop1, RGD1304773
Chromosome Location:
Chr.13: 76942933 - 77076015 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01483613_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rfwd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025126.1 NP_001020297.1 12 - 13 1466 136
XM_006250113.2 XP_006250175.1 19 - 20 2665 136
XM_006250114.2 XP_006250176.1 19 - 20 2653 136
XM_006250115.2 XP_006250177.1 18 - 19 2605 136
XM_006250116.2 XP_006250178.1 18 - 19 2593 136
GenBank mRNA BC091284.1 - 12 - 13 1466 7304516 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300000580.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rfwd2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

response to ionizing radiation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
zinc ion binding
ubiquitin protein ligase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P470775 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 913684 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 639695 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 772076 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P470775 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P470775 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P470775 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 104065 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10940637 - -
Affymetrix Rat Gene 1.1 ST Array 10940637 - -
Rat Genome 230 2.0 Array 1373820_at - -
Rat Genome 230 2.0 Array 1375172_at - -
Rat Genome 230 2.0 Array 1375145_PM_at - -
Rat Genome 230 2.0 Array 1375145_at - -
Rat Genome 230 2.0 Array 1373820_PM_at - -
Rat Genome 230 2.0 Array 1375172_PM_at - -
Affymetrix HT Rat Focus Array 1373820_at - -
Affymetrix HT Rat Focus Array 1373820_PM_at - -
Affymetrix Rat Clariom S Assay PSR1300004024.rn.1 TC1300000580.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300004025.rn.1 TC1300000580.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300004023.rn.1 TC1300000580.rn.1 RTA TC
Rat Expression Set 230 1375172_PM_at - -
Rat Expression Set 230 1375145_PM_at - -
Rat Expression Set 230 1375145_at - -
Rat Expression Set 230 1375172_at - -
Rat Expression Set 230 1373820_PM_at - -
Rat Expression Set 230 1373820_at - -
Affymetrix Rat Gene 1.0 ST Array 10940637 - -
Rat Genome U34 Set rc_AA924670_at - -
Rat Genome U34 Set rc_AA850517_at - -
Rat Genome U34 Set rc_AI230308_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21824750 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1352148 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0