This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01483456_m1
See other ILDR2 GE Assays ›
Gene Symbol:
Ildr2
Gene Name:
immunoglobulin-like domain containing receptor 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 84474191 - 84532347 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01483456_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ildr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008769713.1 XP_008767935.1 8 - 9 1377 61
XM_008769714.1 XP_008767936.1 8 - 9 1316 61
XM_008769715.1 XP_008767937.1 7 - 8 1320 61
XM_008769716.1 XP_008767938.1 8 - 9 3311 61
XM_008769717.1 XP_008767939.1 8 - 9 3289 61
XM_008769718.1 XP_008767940.1 8 - 9 1181 61
XM_008769719.1 XP_008767941.1 7 - 8 1200 61
XM_008769720.1 XP_008767942.1 5 - 6 996 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

response to glucose
insulin secretion
cell differentiation
pancreas development
homeostasis of number of cells within a tissue

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052879 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052879 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052879 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1384035_PM_at - -
Rat Genome 230 2.0 Array 1384035_at - -
Rat Genome 230 2.0 Array 1384036_PM_s_at - -
Rat Genome 230 2.0 Array 1384036_s_at - -
Rat Expression Set 230 1384036_PM_s_at - -
Rat Expression Set 230 1384036_s_at - -
Rat Expression Set 230 1384035_at - -
Rat Expression Set 230 1384035_PM_at - -
Rat Genome U34 Set rc_AA996523_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22132439 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0