This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01481825_m1
See other CABP1 GE Assays ›
Gene Symbol:
Cabp1
Gene Name:
calcium binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 47185449 - 47209785 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01481825_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033675.1 NP_001028847.1 3 - 4 219 97
NM_001033676.1 NP_001028848.1 3 - 4 831 97
NM_133529.2 NP_598213.1 4 - 5 399 97
GenBank mRNA AJ315657.1 - 4 - 5 399 97
AJ315761.1 - 3 - 4 219 97
Y17048.2 - 3 - 4 831 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000782.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cabp1

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
negative regulation of cell communication by electrical coupling
negative regulation of protein import into nucleus
regulation of synaptic plasticity
regulation of voltage-gated calcium channel activity
negative regulation of voltage-gated calcium channel activity
calcium ion binding
protein binding
nuclear localization sequence binding
protein domain specific binding
type 3 metabotropic glutamate receptor binding
protein homodimerization activity
ion channel binding
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200006887.rn.1 TC1200000782.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0