This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01478957_m1
See other FKBP2 GE Assays ›
Gene Symbol:
Fkbp2
Gene Name:
FK506 binding protein 2
Gene Aliases:
RGD1560660
Chromosome Location:
Chr.1: 222232349 - 222235154 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01478957_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fkbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134429.1 NP_001127901.1 1 - 2 89 90
XM_006230756.1 XP_006230818.1 1 - 2 141 90
GenBank mRNA H35224.1 - 1 - 2 43 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001134429.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
FK506 binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100057769.rn.1 TC0100007182.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0