This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01476637_g1
See other IGFLR1 GE Assays ›
Gene Symbol:
Igflr1
Gene Name:
IGF-like family receptor 1
Gene Aliases:
RGD1563574, Tmem149
Chromosome Location:
Chr.1: 89019749 - 89022221 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01476637_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igflr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134610.1 NP_001128082.1 1 - 2 298 136
XM_006228787.2 XP_006228849.1 1 - 2 501 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100001252.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100009767.rn.1 TC0100001252.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100009768.rn.1 TC0100001252.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100009770.rn.1 TC0100001252.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100009772.rn.1 TC0100001252.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0