This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01474550_m1
See other LOC102554620 GE Assays ›
Gene Symbol:
LOC102554620 Vwce
Gene Name:
von Willebrand factor C and EGF domain-containing protein-like
von Willebrand factor C and EGF domains
Gene Aliases:
-
RGD1309910
Chromosome Location:
Chr.Un|NW_007906915: 65166 - 86730 on Build Rnor_6.0
Chromosome Location:
Chr.1: 226687258 - 226717989 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01474550_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102554620
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008773895.1 XP_008772117.1 678 115
XM_008773896.1 XP_008772118.1 6 - 7 678 115
XM_008773897.1 XP_008772119.1 1145 115
XM_008773898.1 XP_008772120.1 6 - 7 678 115
Gene Symbol: Vwce
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271311.1 NP_001258240.1 7 - 8 792 115

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell proliferation
regulation of signal transduction
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0