This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01468750_m1
See other AGR3 GE Assays ›
Gene Symbol:
Agr3
Gene Name:
anterior gradient 3
Gene Aliases:
RGD1565406
Chromosome Location:
Chr.6: 55371738 - 55424233 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01468750_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106724.1 NP_001100194.1 1 - 2 109 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600000620.rn.1

Additional Information:

This assay is in the 5'UTR region of the NM_001106724.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Process(es)

dystroglycan binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600004393.rn.1 TC0600000620.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600004394.rn.1 TC0600000620.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0