This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01468255_m1
See other GPR183 GE Assays ›
Gene Symbol:
Gpr183
Gene Name:
G protein-coupled receptor 183
Gene Aliases:
Ebi2
Chromosome Location:
Chr.15: 108364701 - 108376221 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01468255_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr183
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109386.1 NP_001102856.1 1 - 2 54 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001109386.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

mature B cell differentiation involved in immune response
humoral immune response
G-protein coupled receptor signaling pathway
biological_process
positive regulation of B cell proliferation
positive regulation of ERK1 and ERK2 cascade
molecular_function
G-protein coupled receptor activity
oxysterol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1500016347.rn.1 TC1500002435.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500016348.rn.1 TC1500002435.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0