This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01462185_m1
See other EGFEM1 GE Assays ›
Gene Symbol:
Egfem1
Gene Name:
EGF-like and EMI domain containing 1
Gene Aliases:
RGD1307225
Chromosome Location:
Chr.2: 116526140 - 117152634 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01462185_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Egfem1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107663.1 NP_001101133.1 10 - 11 1460 138
XM_008760923.1 XP_008759145.1 10 - 11 1548 138

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200000942.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Process(es)

calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200005726.rn.1 TC0200000942.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0