This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01461624_m1
See other SLC16A3 GE Assays ›
Gene Symbol:
Slc16a3
Gene Name:
solute carrier family 16 (monocarboxylate transporter), member 3
Gene Aliases:
MCT4, Mct3
Chromosome Location:
Chr.10: 110138523 - 110148360 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01461624_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc16a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030834.1 NP_110461.1 1 - 2 68 85
GenBank mRNA U87627.1 - 1 - 2 68 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_030834.1, U87627.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc16a3

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

plasma membrane lactate transport
lactate transmembrane transporter activity
symporter activity
secondary active monocarboxylate transmembrane transporter activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000019002.rn.1 TC1000002097.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000019009.rn.1 TC1000002097.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0