This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01461204_m1
See other CNP GE Assays ›
Gene Symbol:
Cnp
Gene Name:
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases:
CNPF, CNPI, CNPII, Cnp1
Chromosome Location:
Chr.10: 88489993 - 88497357 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01461204_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012809.2 NP_036941.1 3 - 4 898 106
XM_006247260.2 XP_006247322.1 3 - 4 918 106
XM_006247261.2 XP_006247323.1 3 - 4 922 106
XM_008767948.1 XP_008766170.1 3 - 4 832 106
GenBank mRNA BC098066.1 - 3 - 4 898 7458826 106
L16532.1 - 3 - 4 897 106
M18630.1 - 2 - 3 1191 106

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cnp

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
axonogenesis
aging
adult locomotory behavior
cyclic nucleotide catabolic process
response to toxic substance
substantia nigra development
forebrain development
response to lipopolysaccharide
regulation of mitochondrial membrane permeability
oligodendrocyte differentiation
RNA binding
2',3'-cyclic-nucleotide 3'-phosphodiesterase activity
cyclic nucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0